Learn More
Medchemexpress LLC Thiabendazole-d4 (2-(4-thiazolyl)benzimidazole-d4) | 1190007-20-5 | 99.8% | 1 MG
SDP
Catalog No. 5000405593
Encompass_Preferred
Thiabendazole-d4 is a deuterated form of Thiabendazole, acting as an antiseptic, antifungal, and antiparasitic agent. This compound is primarily used as a tracer and as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-05593 Supplier Medchemexpress LLC Supplier No. HYB0263S1MG
Thiabendazole-d4 is a deuterated form of Thiabendazole, acting as an antiseptic, antifungal, and antiparasitic agent. This compound is primarily used as a tracer and as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.