Learn More
Medchemexpress LLC Thiabendazole (Standard) | 148-79-8 | 99.9% | 250 MG
SDP
Catalog No. 5000406489
Encompass_Preferred
Thiabendazole (Standard) is the analytical standard of Thiabendazole. It is intended for research and analytical applications. Thiabendazole inhibits worm-specific fumarate reductase and has anthelmintic properties. It is for research use only and not sold to patients.
  • Serves as a reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-06489 Supplier Medchemexpress LLC Supplier No. 250MGHYB0263R
Thiabendazole (Standard) is the analytical standard of Thiabendazole. It is intended for research and analytical applications. Thiabendazole inhibits worm-specific fumarate reductase and has anthelmintic properties. It is for research use only and not sold to patients.
  • Serves as a reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS