Learn More
Medchemexpress LLC Thiacetazone | 104-06-3 | 99.7% | 250 MG
SDP
Catalog No. 5000413952
Encompass_Preferred
Thiacetazone (Thioacetazone) is a thiourea-containing antitubercular agent and an orally active antibiotic. It has antibacterial action, inhibiting the growth of *Mycobacterium tuberculosis* H37Rv with a MIC value of 0.1 μg/mL. Thiacetazone is a prodrug activated by the mycobacterial monooxygenase EthA.
  • Inhibits growth of *Mycobacterium tuberculosis* H37Rv.
  • Orally active antibiotic.
  • Activated by mycobacterial monooxygenase EthA.
  • Solid, white to off-white appearance.
Catalog No. 50-004-13952 Supplier Medchemexpress LLC Supplier No. 250MGHYB1526
Thiacetazone (Thioacetazone) is a thiourea-containing antitubercular agent and an orally active antibiotic. It has antibacterial action, inhibiting the growth of *Mycobacterium tuberculosis* H37Rv with a MIC value of 0.1 μg/mL. Thiacetazone is a prodrug activated by the mycobacterial monooxygenase EthA.
  • Inhibits growth of *Mycobacterium tuberculosis* H37Rv.
  • Orally active antibiotic.
  • Activated by mycobacterial monooxygenase EthA.
  • Solid, white to off-white appearance.