Learn More
Medchemexpress LLC Thiacetazone | 104-06-3 | MFCD00022157 | 99.7% | 236.29 | 200 MG
SDP
Catalog No. 5000366042
Encompass_Preferred
Thiacetazone is a thiourea-containing antitubercular agent and an orally active antibiotic. It exhibits antibacterial action by inhibiting the growth of *Mycobacterium tuberculosis* H37Rv with a MIC value of 0.1 μg/mL. This prodrug is activated by the mycobacterial monooxygenase EthA.
  • Thiourea-containing antitubercular agent
  • Orally active antibiotic
  • Inhibits growth of *Mycobacterium tuberculosis* H37Rv
  • Exhibits antibacterial action with a MIC value of 0.1 μg/mL
  • Activated by mycobacterial monooxygenase EthA
Catalog No. 50-003-66042 Supplier Medchemexpress LLC Supplier No. 200MGHYB1526
Thiacetazone is a thiourea-containing antitubercular agent and an orally active antibiotic. It exhibits antibacterial action by inhibiting the growth of *Mycobacterium tuberculosis* H37Rv with a MIC value of 0.1 μg/mL. This prodrug is activated by the mycobacterial monooxygenase EthA.
  • Thiourea-containing antitubercular agent
  • Orally active antibiotic
  • Inhibits growth of *Mycobacterium tuberculosis* H37Rv
  • Exhibits antibacterial action with a MIC value of 0.1 μg/mL
  • Activated by mycobacterial monooxygenase EthA