Learn More
Medchemexpress LLC Thiacetazone (Thioacetazone; Amithiozone) | 104-06-3 | 99.7% | 1 ML
SDP
Catalog No. 5000412029
Encompass_Preferred
Thiacetazone (Thioacetazone) is a thiourea-containing antitubercular agent and an orally active antibiotic. It exhibits antibacterial action, inhibiting the growth of Mycobacterium tuberculosis H37Rv with a MIC value of 0.1 μg/mL. This product is a solution that is ready for reconstitution.
  • Antitubercular agent
  • Orally active antibiotic
  • Exhibits antibacterial action
  • Inhibits growth of Mycobacterium tuberculosis H37Rv
  • Ready for reconstitution solution
Catalog No. 50-004-12029 Supplier Medchemexpress LLC Supplier No. HYB152610MM/1ML
Thiacetazone (Thioacetazone) is a thiourea-containing antitubercular agent and an orally active antibiotic. It exhibits antibacterial action, inhibiting the growth of Mycobacterium tuberculosis H37Rv with a MIC value of 0.1 μg/mL. This product is a solution that is ready for reconstitution.
  • Antitubercular agent
  • Orally active antibiotic
  • Exhibits antibacterial action
  • Inhibits growth of Mycobacterium tuberculosis H37Rv
  • Ready for reconstitution solution