Learn More
Cayman Chemical Thiacloprid, Purity: ≥98%, CAS: 111988-49-9, 50 mg
SDP
Catalog No. 502932757
Encompass_Preferred
A neonicotinoid insecticide; an antagonist of nAChRs (IC50 = 2.7 nM for the D. melanogaster enzyme); lethal to D. melanogaster (LD50 = 0.03 mg/kg); toxic to mice (LD50 = 28 mg/kg)
Catalog No. 50-293-2757 Supplier Cayman Chemical Supplier No. 2582450 MG
A neonicotinoid insecticide; an antagonist of nAChRs (IC50 = 2.7 nM for the D. melanogaster enzyme); lethal to D. melanogaster (LD50 = 0.03 mg/kg); toxic to mice (LD50 = 28 mg/kg)