Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thialysine hydrochloride | 4099-35-8 | 98.0% | 1 G
SDP

Catalog No. 5000230792
Encompass_Preferred

Thialysine hydrochloride is the hydrochloride salt of thialysine, a cysteine-derived amino acid analog used for biochemical and protein-synthesis research.

  • Purity 98.0%
  • Molecular weight 200.69 g/mol
  • Chemical formula C5H13ClN2O2S
  • Appearance white to off-white solid
  • Available pack sizes include 250 MG, 500 MG, 1 G, 5 G, 10 G
  • Store sealed at -20°C; in solvent: -80°C (6 months), -20°C (1 month)
  • Datasheet, COA, and SDS available

Catalog No. 50-002-30792 Supplier Medchemexpress LLC Supplier No. HY1225261G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thialysine hydrochloride is the hydrochloride salt of thialysine, a cysteine-derived amino acid analog used for biochemical and protein-synthesis research.

  • Purity 98.0%
  • Molecular weight 200.69 g/mol
  • Chemical formula C5H13ClN2O2S
  • Appearance white to off-white solid
  • Available pack sizes include 250 MG, 500 MG, 1 G, 5 G, 10 G
  • Store sealed at -20°C; in solvent: -80°C (6 months), -20°C (1 month)
  • Datasheet, COA, and SDS available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.