Learn More
Medchemexpress LLC HY-12588 25mg Medchemexpress, Thiamet G CAS:1009816-48-1 Purity:>98%
SDP
Catalog No. 502028895
Encompass_Preferred
Medchemexpress, HY-12588 25mg Thiamet G CAS:1009816-48-1 Thiamet G is a potent and selective inhibitor of O-GlcNAcase (OGA), which acts to remove O-GlcNAc from modified proteins, with Ki of 20 nM for human OGA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8895 Supplier Medchemexpress LLC Supplier No. HY1258825MG
Medchemexpress, HY-12588 25mg Thiamet G CAS:1009816-48-1 Thiamet G is a potent and selective inhibitor of O-GlcNAcase (OGA), which acts to remove O-GlcNAc from modified proteins, with Ki of 20 nM for human OGA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.