Learn More
TARGETMOL CHEMICALS INC THIAMET G 25MG
SDP
Catalog No. 502579605
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Thiamet G is a potent selective O-GlcNAcase inhibitor with Kiof 21 nM while exhibiting 37 000-fold selectivity over human lysosomal –hexosaminidase. purity: 99%
Catalog No. 50-257-9605 Supplier TARGETMOL CHEMICALS INC Supplier No. T605625MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Thiamet G is a potent selective O-GlcNAcase inhibitor with Kiof 21 nM while exhibiting 37 000-fold selectivity over human lysosomal –hexosaminidase. purity: 99%