Learn More
Chem Service Inc Thiamethoxam Solution, 153719-23-4, MFCD03792862, 1 mL
SDP
Catalog No. NC0556608
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Concentration and Solvent: 100 ug/ml in Methanol, Alternate Name(s): Actara.
Catalog No. NC0556608 Supplier Chem Service Inc Supplier No. S13562M11ML
This item has a minimum order quantity of 2 per supplier requirements
Concentration and Solvent: 100 ug/ml in Methanol, Alternate Name(s): Actara.