Learn More
Cayman Chemical Thiamethoxam, Purity: ≥98%, CAS: 153719-23-4, 50 mg
SDP
Catalog No. 502932778
Encompass_Preferred
A neonicotinoid insecticide and a prodrug form of clothianidin; binds to nAChRs with low affinity (IC50 = 5,000 nM for housefly head membranes) and does not activate non-desensitizing nAChRs in cockroach neurons in vitro; leaf or drench application of thiamethoxam reduces the number of aphids on potato plants in field studies at 100 or 200 mg/L; induces 100% mortality of second instar larvae of the fall armyworm S. frugiperda and the tobacco budworm H. virescens in leaf-dip bioassays at 200 mg/L
Catalog No. 50-293-2778 Supplier Cayman Chemical Supplier No. 2578450 MG
A neonicotinoid insecticide and a prodrug form of clothianidin; binds to nAChRs with low affinity (IC50 = 5,000 nM for housefly head membranes) and does not activate non-desensitizing nAChRs in cockroach neurons in vitro; leaf or drench application of thiamethoxam reduces the number of aphids on potato plants in field studies at 100 or 200 mg/L; induces 100% mortality of second instar larvae of the fall armyworm S. frugiperda and the tobacco budworm H. virescens in leaf-dip bioassays at 200 mg/L