Learn More
AdipoGen Thiamethoxam
SDP
Supplier:  AdipoGen CDXT0114G001
Encompass_Preferred
Chemical. CAS 153719-23-4. Formula C8H10ClN5O3S. MW 291.71. Thiamethoxam is a broad-spectrum, systemic neonicotinoid insecticide, which means it is absorbed quickly by plants and transported to all of its parts, including pollen, where it acts to deter insect feeding. The compound gets in the way of information transfer between nerve cells by interfering with nicotinic acetylcholine receptors in the central nervous system, and eventually paralyzes the muscles of the insects. Compound can be used as analytical reference material.
Catalog No. 50-256-0002