Learn More
Medchemexpress LLC Thiamethoxam-d3 | 1294048-82-0 | 99.0% | 1 MG
SDP
Catalog No. 5000407643
Encompass_Preferred
Thiamethoxam-d3 is the deuterium labeled Thiamethoxam. Thiamethoxam is a broad spectrum neonicotinoid insecticide.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07643 Supplier Medchemexpress LLC Supplier No. HYB0833S11MG
Thiamethoxam-d3 is the deuterium labeled Thiamethoxam. Thiamethoxam is a broad spectrum neonicotinoid insecticide.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.