Learn More
Cayman Chemical Thiamethoxam-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1294048-82-0, 1 mg
SDP
Catalog No. 502912985
Encompass_Preferred
An internal standard for the quantification of thiamethoxam by GC- or LC-MS
Catalog No. 50-291-2985 Supplier Cayman Chemical Supplier No. 356851 MG
An internal standard for the quantification of thiamethoxam by GC- or LC-MS