Learn More
AdipoGen Thiamethoxam-d3
SDP
Supplier:  AdipoGen CDXT0142M025
Encompass_Preferred
Chemical. CAS 1294048-82-0. Formula C8D3H7ClN5O3S. MW 294.73. Thiamethoxam-d3 is deuterium labelled Thiamethoxam, which is a neonicotinoid insecticide. Neonicotinoids NNIs are a family of active substances used in plant protection to control harmful insects and also in veterinary medicine. Neonicotinoids act selectively on the insect nicotinic acetylcholine receptors nAChRs, initially stimulating postsynaptic receptors, increasing Na ingress and K egress and then paralyzing nerve conduction, resulting in rapid death. This compound can be used as a reference compound, in the determination of several neonicotinoid insecticides present in bee pollen, using ultra-high performance liquid chromatography UHPLC coupled with mass spectrometry MS.
Catalog No. 50-255-9986