Learn More
Cayman Chemical Thiamine Pyrophosphate (chloride), Purity: ≥95%, CAS: 154-87-0, 1 g
SDP
Catalog No. 502982883
Encompass_Preferred
A metabolite of vitamin B1; a cofactor synthesized in the cytosol; required for the activity of cytosolic transketolase and mitochondrial pyruvate, oxoglutarate, and branched-chain keto acid dehydrogenases; prevents hyperglycemia-induced retinopathy and desflurane-induced hepatotoxicity in rats
Catalog No. 50-298-2883 Supplier Cayman Chemical Supplier No. 202541 G
A metabolite of vitamin B1; a cofactor synthesized in the cytosol; required for the activity of cytosolic transketolase and mitochondrial pyruvate, oxoglutarate, and branched-chain keto acid dehydrogenases; prevents hyperglycemia-induced retinopathy and desflurane-induced hepatotoxicity in rats