Learn More
Medchemexpress LLC Thiamine-13C3 hydrochloride | 340.25 | 1 MG
SDP
Catalog No. 5000416708
Encompass_Preferred
Thiamine-13C3 hydrochloride is the 13C-labeled version of Thiamine hydrochloride, an essential micronutrient and a cofactor for numerous central metabolic enzymes. Intended for research use only, this compound is not sold to patients. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Labeled with stable heavy isotopes for quantitation in drug development
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
Catalog No. 50-004-16708 Supplier Medchemexpress LLC Supplier No. HYN0680S31MG
Thiamine-13C3 hydrochloride is the 13C-labeled version of Thiamine hydrochloride, an essential micronutrient and a cofactor for numerous central metabolic enzymes. Intended for research use only, this compound is not sold to patients. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Labeled with stable heavy isotopes for quantitation in drug development
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications