Learn More
American Radiolabeled Chemicals Inc Thiamine [3H(G)] Hydrochloride, 50 µCi
SDP
Catalog No. NC1509241
Encompass_Preferred
Molecular Weight: 337.3, Concentration: 1 mCi/ml, Specific Activity: 10-20 Ci/mmol 370-740 GBq/mmol
Catalog No. NC1509241 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 071050UCI
Molecular Weight: 337.3, Concentration: 1 mCi/ml, Specific Activity: 10-20 Ci/mmol 370-740 GBq/mmol