Learn More
Medchemexpress LLC Thiamine-d4 hydrochloride | 67-03-8 | 341.29 | 1 MG
SDP
Catalog No. 5000415833
Encompass_Preferred
Thiamine-d4 hydrochloride is a deuterium labeled form of Thiamine hydrochloride, an essential micronutrient and cofactor for many central metabolic enzymes. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-15833 Supplier Medchemexpress LLC Supplier No. HYN0680S21MG
Thiamine-d4 hydrochloride is a deuterium labeled form of Thiamine hydrochloride, an essential micronutrient and cofactor for many central metabolic enzymes. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles