Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Disulfide Hydrate 98.0+%, TCI America™
SDP

Catalog No. T017625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T017625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T017625G Supplier TCI America Supplier No. T017625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-16-3
Molecular Formula C24H34N8O4S2
Molecular Weight (g/mol) 562.708
MDL Number MFCD00010427
InChI Key GFEGEDUIIYDMOX-KBNZVFGVSA-N
Synonym Aneurine Disulfide
PubChem CID 3085168
IUPAC Name N-[(4-amino-2-methylpyrimidin-5-yl)methyl]-N-[(Z)-3-[[(E)-2-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-5-hydroxypent-2-en-3-yl]disulfanyl]-5-hydroxypent-2-en-2-yl]formamide
SMILES CC1=NC=C(C(=N1)N)CN(C=O)C(=C(CCO)SSC(=C(C)N(CC2=CN=C(N=C2N)C)C=O)CCO)C

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 562.71
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiamine Disulfide Hydrate
EINECSNumber (9)-0812
RTECSNumber LQ2715000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.