Learn More
U.S. Pharmacopeia Thiamine Hydrochloride, 67-03-8, MFCD00012780, 500mg
Catalog No. NC9612355
Encompass
Molecular formula C12H17ClN4OS . HCl, Molecular weight 337.3, Melting Point 478.4 °F (248 °C) (decomposes), Synonyms: Vitamin B1 hydrochloride
Catalog No. NC9612355 Supplier U.S. Pharmacopeia Supplier No. 1656002
Molecular formula C12H17ClN4OS . HCl, Molecular weight 337.3, Melting Point 478.4 °F (248 °C) (decomposes), Synonyms: Vitamin B1 hydrochloride