Learn More
Sigma Aldrich Fine Chemicals Biosciences Thiamine Hydrochloride | CAS 67-03-8 | 1 kg
Catalog No. NC2082497
Encompass
Thiamine hydrochloride, also known as vitamin B1, is an essential water-soluble vitamin that is commonly used to prevent and treat thiamine deficiency. It plays an important role in the breakdown of carbohydrates and branched chain amino acids in cellular metabolism. Thiamine hydrochloride | Purity: ≥99% | Mol Wt: 337.27 | CAS No: 67-03-8 | Unit of Measure: 1 kg
Catalog No. NC2082497 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 47858
Thiamine hydrochloride, also known as vitamin B1, is an essential water-soluble vitamin that is commonly used to prevent and treat thiamine deficiency. It plays an important role in the breakdown of carbohydrates and branched chain amino acids in cellular metabolism. Thiamine hydrochloride | Purity: ≥99% | Mol Wt: 337.27 | CAS No: 67-03-8 | Unit of Measure: 1 kg