Learn More
Sigma Aldrich Fine Chemicals Biosciences Thiamine hydrochloride | 67-03-8 | MFCD00012780 | 100g
Catalog No. NC1592514
Encompass
Thiamine hydrochloride | Purity: ≥99% | MW: 337.27 | 67-03-8 | MFCD00012780 | 100g
Catalog No. NC1592514 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T4625100G
Thiamine hydrochloride | Purity: ≥99% | MW: 337.27 | 67-03-8 | MFCD00012780 | 100g