Learn More
Medchemexpress LLC Thiamine hydrochloride | 67-03-8 | 337.27 | 1 ML
SDP
Catalog No. 5000388673
Encompass_Preferred
Thiamine hydrochloride is an essential micronutrient serving as a cofactor for many central metabolic enzymes. It activates NrF-2/HO-1 and inhibits TLR4, NF-κB, exhibiting neuroprotective and anti-inflammatory effects. It is utilized in studies concerning diabetic complications, neurological diseases, cancers, and colitis.
  • Essential micronutrient and cofactor for central metabolic enzymes.
  • Activates NrF-2/HO-1 and inhibits TLR4, NF-κB.
  • Exhibits neuroprotective and anti-inflammatory effects.
  • Used in studies of diabetic complications, neurological diseases, cancers, and colitis.
  • Increases cellular PDH activities and oxygen consumption rate in breast cancer cells.
  • Reduces proliferation in cancer cell lines.
  • Ameliorates scopolamine-induced memory dysfunction in adult mice.
  • Demonstrates anti-inflammatory and antihyperalgesic effects in adjuvant-induced arthritis rats.
Catalog No. 50-003-88673 Supplier Medchemexpress LLC Supplier No. HYN068010MM/1ML
Thiamine hydrochloride is an essential micronutrient serving as a cofactor for many central metabolic enzymes. It activates NrF-2/HO-1 and inhibits TLR4, NF-κB, exhibiting neuroprotective and anti-inflammatory effects. It is utilized in studies concerning diabetic complications, neurological diseases, cancers, and colitis.
  • Essential micronutrient and cofactor for central metabolic enzymes.
  • Activates NrF-2/HO-1 and inhibits TLR4, NF-κB.
  • Exhibits neuroprotective and anti-inflammatory effects.
  • Used in studies of diabetic complications, neurological diseases, cancers, and colitis.
  • Increases cellular PDH activities and oxygen consumption rate in breast cancer cells.
  • Reduces proliferation in cancer cell lines.
  • Ameliorates scopolamine-induced memory dysfunction in adult mice.
  • Demonstrates anti-inflammatory and antihyperalgesic effects in adjuvant-induced arthritis rats.