Learn More
Medchemexpress LLC Thiamine hydrochloride | 67-03-8 | 337.27 | 100 MG
SDP
Catalog No. 5000388830
Encompass_Preferred
Thiamine hydrochloride is an essential micronutrient that serves as a cofactor for numerous central metabolic enzymes. It activates NrF-2/HO-1 and inhibits TLR4, NF-κB, providing neuroprotective and anti-inflammatory effects. This compound is utilized in research for diabetic complications, neurological diseases, cancers, and colitis.
  • Essential micronutrient and cofactor for central metabolic enzymes
  • Activates NrF-2/HO-1 and inhibits TLR4, NF-κB
  • Neuroprotective effects
  • Anti-inflammatory effects
  • Used in studies of diabetic complications, neurological diseases, cancers, and colitis
Catalog No. 50-003-88830 Supplier Medchemexpress LLC Supplier No. HYN0680100MG
Thiamine hydrochloride is an essential micronutrient that serves as a cofactor for numerous central metabolic enzymes. It activates NrF-2/HO-1 and inhibits TLR4, NF-κB, providing neuroprotective and anti-inflammatory effects. This compound is utilized in research for diabetic complications, neurological diseases, cancers, and colitis.
  • Essential micronutrient and cofactor for central metabolic enzymes
  • Activates NrF-2/HO-1 and inhibits TLR4, NF-κB
  • Neuroprotective effects
  • Anti-inflammatory effects
  • Used in studies of diabetic complications, neurological diseases, cancers, and colitis