Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Hydrochloride (B1), MilliporeSigma™ Supelco™

Catalog No. 111015126 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5126 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-5126 Supplier MilliporeSigma Supelco Supplier No. 47858
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thiamine hydrochloride (B1) is a water soluble vitamin, which belongs the B-complex group of vitamins. It can exist both as a hemihydrate and as a nonstoichiometric hydrate. It can generally act like a co-enzyme for biochemical reactions involved in carbohydrate metabolism.

Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 67-03-8
Physical Form Neat
Linear Formula C12H17 ClN4 OS · HCl
MDL Number MFCD00012780
Melting Point 250°C
Synonym Thiamine hydrochloride; Aneurine hydrochloride; Vitamin B1 hydrochloride
Type Analytical Standard
Quantity 1000 mg
Formula Weight 337.27
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.