Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiamine monochloride-13C4 hydrochloride | 1257525-77-1 | 341.24 | 1 MG
SDP

Catalog No. 5000388880
Encompass_Preferred

Thiamine monochloride-13C4 (hydrochloride) is the deuterium labeled Thiamine hydrochloride. Thiamine hydrochloride is an essential micronutrient needed as a cofactor for many central metabolic enzymes.

  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity: 98.39%
  • Appearance: Solid, white to off-white
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs

Catalog No. 50-003-88880 Supplier Medchemexpress LLC Supplier No. HYN0680S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiamine monochloride-13C4 (hydrochloride) is the deuterium labeled Thiamine hydrochloride. Thiamine hydrochloride is an essential micronutrient needed as a cofactor for many central metabolic enzymes.

  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity: 98.39%
  • Appearance: Solid, white to off-white
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.