Learn More
Medchemexpress LLC HY-A0100 500mg Medchemexpress, Thiamine monochloride CAS:59-43-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0100500MG
Encompass_Preferred
Medchemexpress, HY-A0100 500mg Thiamine monochloride CAS:59-43-8 Thiamine monochloride (Vitamin B1) is an essential vitamin that plays an important role in cellular production of energy from ingested food and enhances normal neuronal actives. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2325