Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiamine monophosphate chloride | 532-40-1 | MFCD00063473 | 99.3% | 380.79 g/mol | C12H18ClN4O4PS | 10 MG
SDP

Catalog No. 5000298107
Encompass_Preferred

Thiamine monophosphate chloride is a water-soluble derivative of vitamin B1 used as a source of thiamine in nutritional supplements and as a research reagent for studies of supplementation, formulation, and metabolism.

  • Water-soluble source of vitamin B1 suitable for supplement formulation and nutritional research.
  • Molecular formula C12H18ClN4O4PS and molecular weight 380.79 g/mol.
  • CAS number 532-40-1 for definitive chemical identification.
  • Available in small research pack sizes, such as 5 mg, 10 mg, 50 mg, and 100 mg.
  • High purity suitable for analytical and formulation work (purity reported ~99.3%).

Catalog No. 50-002-98107 Supplier Medchemexpress LLC Supplier No. HYB170110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiamine monophosphate chloride is a water-soluble derivative of vitamin B1 used as a source of thiamine in nutritional supplements and as a research reagent for studies of supplementation, formulation, and metabolism.

  • Water-soluble source of vitamin B1 suitable for supplement formulation and nutritional research.
  • Molecular formula C12H18ClN4O4PS and molecular weight 380.79 g/mol.
  • CAS number 532-40-1 for definitive chemical identification.
  • Available in small research pack sizes, such as 5 mg, 10 mg, 50 mg, and 100 mg.
  • High purity suitable for analytical and formulation work (purity reported ~99.3%).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.