Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiamine monophosphate chloride (dihydrate) | 273724-21-3 | 99.2% | 416.82 | 500 MG
SDP

Catalog No. 5000365845
Encompass_Preferred

Thiamine monophosphate chloride dihydrate is a vitamin B1 metabolite and a thiamine precursor. It is absorbable in vivo and can be converted into thiamine. This compound can also be utilized as a food additive.

  • Exhibits toxicity in rats at certain dosages.
  • Appearance: solid.
  • Color: white to off-white.
  • Initial source: endogenous metabolite.
  • Storage: 4°C, sealed, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month.

Catalog No. 50-003-65845 Supplier Medchemexpress LLC Supplier No. 500MGHY128742
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiamine monophosphate chloride dihydrate is a vitamin B1 metabolite and a thiamine precursor. It is absorbable in vivo and can be converted into thiamine. This compound can also be utilized as a food additive.

  • Exhibits toxicity in rats at certain dosages.
  • Appearance: solid.
  • Color: white to off-white.
  • Initial source: endogenous metabolite.
  • Storage: 4°C, sealed, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.