Learn More
Medchemexpress LLC Thiamine monophosphate chloride dihydrate | 273724-21-3 | 99.24% | 1 ML
SDP
Catalog No. 5000342860
Encompass_Preferred
Thiamine monophosphate chloride dihydrate is a vitamin B1 metabolite and thiamine precursor. It is absorbable in vivo and can be converted into thiamine. It can also be used as a food additive.
  • Vitamin B1 metabolite and thiamine precursor
  • Absorbable in vivo and convertible into thiamine
  • Can be used as a food additive
Catalog No. 50-003-42860 Supplier Medchemexpress LLC Supplier No. HY12874210MM/1ML
Thiamine monophosphate chloride dihydrate is a vitamin B1 metabolite and thiamine precursor. It is absorbable in vivo and can be converted into thiamine. It can also be used as a food additive.
  • Vitamin B1 metabolite and thiamine precursor
  • Absorbable in vivo and convertible into thiamine
  • Can be used as a food additive