Learn More
Apexbio Technology LLC THIAMINE-NITRATE-1G
SDP
Catalog No. 5000568382
Encompass_Preferred
Thiamine nitrate (CAS No 532-43-4) is primarily utilized in biomedical research focusing on vitamin metabolism and mechanisms of action within the human body Originating as a synthetic derivative of thiamine it acts by participating in several crucial metabolic pathways particularly as a cofactor for enzymes involved in carbohydrate metabolism In vitro its activity generally falls within the nanomolar to low micromolar range demonstrating its potency in biochemical assays Common applications include studies involving cell models such as neuronal or cardiac cells and animal studies that aim to understand metabolic disorders or neuropathologies Additionally it is used in drug screening processes to explore potential therapeutic interventions for thiamine deficiency-related conditions Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68382 Supplier Apexbio Technology LLC Supplier No. C69631000
Thiamine nitrate (CAS No 532-43-4) is primarily utilized in biomedical research focusing on vitamin metabolism and mechanisms of action within the human body Originating as a synthetic derivative of thiamine it acts by participating in several crucial metabolic pathways particularly as a cofactor for enzymes involved in carbohydrate metabolism In vitro its activity generally falls within the nanomolar to low micromolar range demonstrating its potency in biochemical assays Common applications include studies involving cell models such as neuronal or cardiac cells and animal studies that aim to understand metabolic disorders or neuropathologies Additionally it is used in drug screening processes to explore potential therapeutic interventions for thiamine deficiency-related conditions Experimental concentrations typically depend on the specific research design