Learn More
Medchemexpress LLC HY-113076 100mg Medchemexpress, Thiamine pyrophosphate CAS:154-87-0 Purity:>98%
SDP
Catalog No. 501932452
Encompass_Preferred
Medchemexpress, HY-113076 100mg Thiamine pyrophosphate CAS:154-87-0 Thiamine pyrophosphate is the coenzyme form of Vitamin B1 and is a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2452 Supplier Medchemexpress LLC Supplier No. HY113076100MG
Medchemexpress, HY-113076 100mg Thiamine pyrophosphate CAS:154-87-0 Thiamine pyrophosphate is the coenzyme form of Vitamin B1 and is a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.