Learn More
Medchemexpress LLC Thiamine pyrophosphate | 154-87-0 | 98.4% | 460.77 | 50 MG
SDP
Catalog No. 5000437730
Encompass_Preferred
Thiamine pyrophosphate is an analytical standard intended for research and analytical applications. It is the coenzyme form of Vitamin B1 and a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. This compound is necessary for oxidative phosphorylation and the pentose phosphate pathway by acting as a cofactor for α-ketoacid dehydrogenases.
  • Used for research and analytical applications
  • Coenzyme form of Vitamin B1
  • Required intermediate in metabolic complexes
  • Necessary for oxidative phosphorylation and the pentose phosphate pathway
  • Acts as a cofactor for α-ketoacid dehydrogenases
  • Analytical standard grade
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-37730 Supplier Medchemexpress LLC Supplier No. HY113076R50MG
Thiamine pyrophosphate is an analytical standard intended for research and analytical applications. It is the coenzyme form of Vitamin B1 and a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. This compound is necessary for oxidative phosphorylation and the pentose phosphate pathway by acting as a cofactor for α-ketoacid dehydrogenases.
  • Used for research and analytical applications
  • Coenzyme form of Vitamin B1
  • Required intermediate in metabolic complexes
  • Necessary for oxidative phosphorylation and the pentose phosphate pathway
  • Acts as a cofactor for α-ketoacid dehydrogenases
  • Analytical standard grade
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS