Learn More
Medchemexpress LLC Thiamine pyrophosphate | 154-87-0 | 99.9% | 460.77 | 1 MG
SDP
Catalog No. 5000438408
Encompass_Preferred
Thiamine pyrophosphate, also known as Cocarboxylase, is the coenzyme form of Vitamin B1. It is a crucial intermediate in the pyruvate dehydrogenase and ketoglutarate dehydrogenase complexes, essential for oxidative phosphorylation and the pentose phosphate pathway. This product acts as a cofactor for α-ketoacid dehydrogenases.
  • Stable under recommended storage conditions.
  • Identified for use as laboratory chemicals and in the manufacture of substances.
  • For research use only.
Catalog No. 50-004-38408 Supplier Medchemexpress LLC Supplier No. HY113076S1MG
Thiamine pyrophosphate, also known as Cocarboxylase, is the coenzyme form of Vitamin B1. It is a crucial intermediate in the pyruvate dehydrogenase and ketoglutarate dehydrogenase complexes, essential for oxidative phosphorylation and the pentose phosphate pathway. This product acts as a cofactor for α-ketoacid dehydrogenases.
  • Stable under recommended storage conditions.
  • Identified for use as laboratory chemicals and in the manufacture of substances.
  • For research use only.