Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Pyrophosphate Chloride, Spectrum™ Chemical
SDP

Catalog No. 18621567 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-567 5 kg
18-621-563 -
18-621-564 -
18-621-565 -
18-621-566 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-621-567 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T15865KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Cocarboxylase

Specifications

Packaging Drum
Quantity 5 kg
Percent Purity 97%
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Thiamine Pyrophosphate Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.