Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Pyrophosphate Chloride 98.0+%, TCI America™
SDP

Catalog No. T01835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T01835G Supplier TCI America Supplier No. T01835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 154-87-0
Color White
Molecular Formula C12H19ClN4O7P2S
Quantity 5 g
MDL Number MFCD00038740
Synonym thiamine pyrophosphate, cocarboxylase, thiamine pyrophosphate chloride, cocarboxilasa, cocarboxylasum, farmaneurina, pyruvodehydrase, berolase, biosyth, bioxylasi
InChI Key YXVCLPJQTZXJLH-UHFFFAOYNA-N
SMILES [Cl-].CC1=C(CCOP(O)(=O)OP(O)(O)=O)SC=[N+]1CC1=CN=C(C)N=C1N
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-{[hydroxy(phosphonooxy)phosphoryl]oxy}ethyl)-4-methyl-1,3-thiazol-3-ium chloride
Molecular Weight (g/mol) 460.76
PubChem CID 9068
ChEBI CHEBI:18290
Formula Weight 460.76
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiamine Pyrophosphate Chloride
Show More Show Less
RTECSNumber XI7552000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.