Learn More
Medchemexpress LLC Thiamine pyrophosphate (Cocarboxylase) | 154-87-0 | 99.9% | 460.77 | 25 MG
SDP
Catalog No. 5000439818
Encompass_Preferred
Thiamine pyrophosphate is the coenzyme form of Vitamin B1, serving as an essential intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. It acts as a cofactor for α-ketoacid dehydrogenases, crucial for oxidative phosphorylation and the pentose phosphate pathway. This product is intended for research use only, not for patient sales, and appears as a solid, white to off-white substance. It is identified as an endogenous metabolite and a human gut microbiota metabolite.
  • Inhibits oxidative damage in rat ovarian tissue.
  • Protects against Cisplatin-induced cardiotoxicity, associated oxidative stress, and heart injury in rats.
Catalog No. 50-004-39818 Supplier Medchemexpress LLC Supplier No. 25MGHY113076R
Thiamine pyrophosphate is the coenzyme form of Vitamin B1, serving as an essential intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. It acts as a cofactor for α-ketoacid dehydrogenases, crucial for oxidative phosphorylation and the pentose phosphate pathway. This product is intended for research use only, not for patient sales, and appears as a solid, white to off-white substance. It is identified as an endogenous metabolite and a human gut microbiota metabolite.
  • Inhibits oxidative damage in rat ovarian tissue.
  • Protects against Cisplatin-induced cardiotoxicity, associated oxidative stress, and heart injury in rats.