Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine pyrophosphate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013237 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3237 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3237 Supplier MilliporeSigma Supelco Supplier No. PHR1369500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Thiamine pyrophosphate is a coenzyme of transketolase that catalyzes the cleavage of ribulose-5-phosphate; thereby forming D-glyceraldehyde-3-phosphate. This reaction requires the addition of an acceptor aldehyde such as ribose-5-phosphate, glyceraldehye or glycolaldehyde.
TRUSTED_SUSTAINABILITY

Specifications

CAS 154-87-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C12H19 ClN4O7P2 S
MDL Number MFCD00038740
Synonym Thiamine pyrophosphate; Aneurinepyrophosphoric acid; Cocarboxylase; Thiamine pyrophosphate chloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 460.77
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.