Learn More
Medchemexpress LLC Thiamine pyrophosphate (Standard) | 154-87-0 | 98.4% | 460.77 | 100 MG
SDP
Catalog No. 5000435600
Encompass_Preferred
Thiamine pyrophosphate (Standard) is the analytical standard of Thiamine pyrophosphate, intended for research and analytical applications. It is the coenzyme form of Vitamin B1 and a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. Thiamine pyrophosphate is necessary for oxidative phosphorylation and the pentose phosphate pathway by acting as a cofactor for α-ketoacid dehydrogenases.
  • Analytical standard of thiamine pyrophosphate
  • Intended for research and analytical applications
  • Coenzyme form of vitamin B1
  • Required intermediate in pyruvate dehydrogenase complex and ketoglutarate dehydrogenase complex
  • Necessary for oxidative phosphorylation and the pentose phosphate pathway
  • Acts as a cofactor for α-ketoacid dehydrogenases
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-35600 Supplier Medchemexpress LLC Supplier No. HY113076R100MG
Thiamine pyrophosphate (Standard) is the analytical standard of Thiamine pyrophosphate, intended for research and analytical applications. It is the coenzyme form of Vitamin B1 and a required intermediate in the pyruvate dehydrogenase complex and the ketoglutarate dehydrogenase complex. Thiamine pyrophosphate is necessary for oxidative phosphorylation and the pentose phosphate pathway by acting as a cofactor for α-ketoacid dehydrogenases.
  • Analytical standard of thiamine pyrophosphate
  • Intended for research and analytical applications
  • Coenzyme form of vitamin B1
  • Required intermediate in pyruvate dehydrogenase complex and ketoglutarate dehydrogenase complex
  • Necessary for oxidative phosphorylation and the pentose phosphate pathway
  • Acts as a cofactor for α-ketoacid dehydrogenases
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS