Learn More
Cayman Chemical Thiamine (nitrate), Purity: ≥98%, CAS: 532-43-4, 10 g
SDP
Catalog No. 502932482
Encompass_Preferred
A water-soluble vitamin with diverse biological activities; inhibits lipid peroxidation in rat liver microsomes and free radical oxidation of oleic acid in vitro from 1-100 μM; reduces hepatic levels of AST and ALT, lipid peroxidation, and protein damage in a rat model of acute ethanol intoxication at 100 mg/kg; reverses predator stress-induced suppression of hippocampal neurogenesis and decreases latency of step-down from a platform in mice; reduces neurodegeneration and increases survival in Slc19a3-/- mice at 8.5 mg/100 g food
Catalog No. 50-293-2482 Supplier Cayman Chemical Supplier No. 2533210 G
A water-soluble vitamin with diverse biological activities; inhibits lipid peroxidation in rat liver microsomes and free radical oxidation of oleic acid in vitro from 1-100 μM; reduces hepatic levels of AST and ALT, lipid peroxidation, and protein damage in a rat model of acute ethanol intoxication at 100 mg/kg; reverses predator stress-induced suppression of hippocampal neurogenesis and decreases latency of step-down from a platform in mice; reduces neurodegeneration and increases survival in Slc19a3-/- mice at 8.5 mg/100 g food