Learn More
Medchemexpress LLC HY-B0479 100mg Medchemexpress, Thiamphenicol CAS:15318-45-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0479100MG
Encompass_Preferred
Medchemexpress, HY-B0479 100mg Thiamphenicol CAS:15318-45-3 Thiamphenicol (Thiophenicol), a methyl-sulfonyl derivative of Chloramphenicol, is a broad-spectrum antimicrobial antibiotic. Thiamphenicol acts by binding to the 50S ribosomal subunit, leading to inhibition of protein synthesis and bacteriostatic effect (against Gram-negative, Gram-positive aerobic and anaerobic bacteria). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2554