Learn More
Cayman Chemical Thiamphenicol, Purity: ≥98%, CAS: 15318-45-3, 10 g
SDP
Catalog No. 502985314
Encompass_Preferred
An antibiotic derived from chloramphenicol that displays a similar spectrum of activity with enhanced potency
Catalog No. 50-298-5314 Supplier Cayman Chemical Supplier No. 2135710 G
An antibiotic derived from chloramphenicol that displays a similar spectrum of activity with enhanced potency