Learn More
Medchemexpress LLC Thiamphenicol | 15318-45-3 | 99.6% | 1 ML
SDP
Catalog No. 5000409209
Encompass_Preferred
Thiamphenicol is a methyl-sulfonyl derivative of Chloramphenicol, acting as a broad-spectrum antimicrobial antibiotic. It inhibits protein synthesis and provides a bacteriostatic effect by binding to the 50S ribosomal subunit, making it effective against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. Studies have indicated rapid absorption and high bioavailability (over 70%) after oral administration.
  • Inhibits protein synthesis
  • Bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria
  • Significant post-antibiotic effect (PAE)
  • Potent bactericidal activity against H. influenzae
  • Good in vitro activity against multi-resistant pathogens
  • Rapid absorption and high bioavailability in vivo
Catalog No. 50-004-09209 Supplier Medchemexpress LLC Supplier No. HYB047910MM/1ML
Thiamphenicol is a methyl-sulfonyl derivative of Chloramphenicol, acting as a broad-spectrum antimicrobial antibiotic. It inhibits protein synthesis and provides a bacteriostatic effect by binding to the 50S ribosomal subunit, making it effective against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. Studies have indicated rapid absorption and high bioavailability (over 70%) after oral administration.
  • Inhibits protein synthesis
  • Bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria
  • Significant post-antibiotic effect (PAE)
  • Potent bactericidal activity against H. influenzae
  • Good in vitro activity against multi-resistant pathogens
  • Rapid absorption and high bioavailability in vivo