Learn More
Medchemexpress LLC Thiamphenicol | 15318-45-3 | 99.6% | 100 MG
SDP
Catalog No. 5000405527
Encompass_Preferred
Thiamphenicol is a broad-spectrum antimicrobial antibiotic, a methyl-sulfonyl derivative of Chloramphenicol. It inhibits protein synthesis by binding to the 50S ribosomal subunit, exerting a bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. This compound is a white to off-white solid intended for research use only.
  • Broad-spectrum antimicrobial antibiotic
  • Methyl-sulfonyl derivative of Chloramphenicol
  • Inhibits protein synthesis by binding to 50S ribosomal subunit
  • Bacteriostatic effect against Gram-negative, Gram-positive, aerobic, and anaerobic bacteria
  • Exhibits significant post-antibiotic effect (PAE)
  • Powerful bactericidal effect against beta-lactamase-positive and -negative H. influenzae
  • Good in vitro activity against difficult-to-treat multiply resistant pathogens
Catalog No. 50-004-05527 Supplier Medchemexpress LLC Supplier No. HYB0479R100MG
Thiamphenicol is a broad-spectrum antimicrobial antibiotic, a methyl-sulfonyl derivative of Chloramphenicol. It inhibits protein synthesis by binding to the 50S ribosomal subunit, exerting a bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. This compound is a white to off-white solid intended for research use only.
  • Broad-spectrum antimicrobial antibiotic
  • Methyl-sulfonyl derivative of Chloramphenicol
  • Inhibits protein synthesis by binding to 50S ribosomal subunit
  • Bacteriostatic effect against Gram-negative, Gram-positive, aerobic, and anaerobic bacteria
  • Exhibits significant post-antibiotic effect (PAE)
  • Powerful bactericidal effect against beta-lactamase-positive and -negative H. influenzae
  • Good in vitro activity against difficult-to-treat multiply resistant pathogens