Learn More
Medchemexpress LLC Thiamphenicol | 15318-45-3 | 99.6% | 500 MG
SDP
Catalog No. 5000409208
Encompass_Preferred
Thiamphenicol is a methyl-sulfonyl derivative of Chloramphenicol and a broad-spectrum antimicrobial antibiotic. It functions by binding to the 50S ribosomal subunit, which inhibits protein synthesis and results in a bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. It is for research use only and not sold to patients.
  • Significant post-antibiotic effect (PAE) on S. pneumoniae, S. aureus, and Escherichia coli.
  • Powerful bactericidal effect against β-lactamase-positive and -negative H. influenzae.
  • Good in vitro activity against difficult-to-treat multiply resistant pathogens.
  • Rapid absorption and bioavailability over 70% (in vivo).
Catalog No. 50-004-09208 Supplier Medchemexpress LLC Supplier No. HYB0479500MG
Thiamphenicol is a methyl-sulfonyl derivative of Chloramphenicol and a broad-spectrum antimicrobial antibiotic. It functions by binding to the 50S ribosomal subunit, which inhibits protein synthesis and results in a bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. It is for research use only and not sold to patients.
  • Significant post-antibiotic effect (PAE) on S. pneumoniae, S. aureus, and Escherichia coli.
  • Powerful bactericidal effect against β-lactamase-positive and -negative H. influenzae.
  • Good in vitro activity against difficult-to-treat multiply resistant pathogens.
  • Rapid absorption and bioavailability over 70% (in vivo).