Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiamphenicol | 15318-45-3 | MFCD00467983 | 99.8% | 356.22 | C12H15Cl2NO5S | 10 MG
SDP

Catalog No. 5000297103
Encompass_Preferred

Thiamphenicol analytical standard for use as a reference material in chromatographic and analytical method development, calibration, and quality control. Supplied as a solid in small package sizes for laboratory use.

  • Analytical standard for thiamphenicol assays and method development.
  • High purity (99.8%) for reliable quantitation and QC.
  • Molecular formula C12H15Cl2NO5S; molecular weight 356.22 g/mol.
  • Available in small package size (10 MG) suitable for analytical use.
  • CAS number 15318-45-3 included for unambiguous identification.
  • Store under conditions specified on the certificate of analysis.

Catalog No. 50-002-97103 Supplier Medchemexpress LLC Supplier No. HYB0479R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiamphenicol analytical standard for use as a reference material in chromatographic and analytical method development, calibration, and quality control. Supplied as a solid in small package sizes for laboratory use.

  • Analytical standard for thiamphenicol assays and method development.
  • High purity (99.8%) for reliable quantitation and QC.
  • Molecular formula C12H15Cl2NO5S; molecular weight 356.22 g/mol.
  • Available in small package size (10 MG) suitable for analytical use.
  • CAS number 15318-45-3 included for unambiguous identification.
  • Store under conditions specified on the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.