Learn More
Selleck Chemical LLC Thiamphenicol S2583-1g
SDP
Catalog No. 503111646
Encompass_Preferred
Thiamphenicol (Thiophenicol Dextrosulphenidol) is an antimicrobial antibiotic and a methyl-sulfonyl analogue of chloramphenicol
Catalog No. 50-311-1646 Supplier Selleck Chemical LLC Supplier No. S25831G
Thiamphenicol (Thiophenicol Dextrosulphenidol) is an antimicrobial antibiotic and a methyl-sulfonyl analogue of chloramphenicol