Learn More
Selleck Chemical LLC Thiamphenicol S2583-50mg
SDP
Catalog No. 503112405
Encompass_Preferred
Thiamphenicol (Thiophenicol Dextrosulphenidol) is an antimicrobial antibiotic and a methyl-sulfonyl analogue of chloramphenicol
Catalog No. 50-311-2405 Supplier Selleck Chemical LLC Supplier No. S258350MG
Thiamphenicol (Thiophenicol Dextrosulphenidol) is an antimicrobial antibiotic and a methyl-sulfonyl analogue of chloramphenicol