Learn More
Alkali Scientific Thiamphenicol, 5 G
SDP
Catalog No. 5000659662
Encompass_Preferred
Thiamphenicol, in its 5 g powder form, is used in laboratory research and clinical settings as a protein synthesis inhibitor. It is effective against a wide range of bacterial infections, particularly those caused by Gram-negative and Gram-positive organisms. Thiamphenicol works by inhibiting bacterial ribosomal protein synthesis, similar to other chloramphenicol-related antibiotics. This 5 g quantity is typically used to prepare larger volumes of solution for clinical treatment or for experimental studies in microbiology, such as antibiotic susceptibility testing and bacterial growth analysis. It is a valuable tool in the study of bacterial resistance mechanisms and therapeutic interventions.
Catalog No. 50-006-59662 Supplier Alkali Scientific Supplier No. HIA002CP
Thiamphenicol, in its 5 g powder form, is used in laboratory research and clinical settings as a protein synthesis inhibitor. It is effective against a wide range of bacterial infections, particularly those caused by Gram-negative and Gram-positive organisms. Thiamphenicol works by inhibiting bacterial ribosomal protein synthesis, similar to other chloramphenicol-related antibiotics. This 5 g quantity is typically used to prepare larger volumes of solution for clinical treatment or for experimental studies in microbiology, such as antibiotic susceptibility testing and bacterial growth analysis. It is a valuable tool in the study of bacterial resistance mechanisms and therapeutic interventions.